CytoSelect™ Cell Migration (Chemotaxis) Assay Kits, 8 µm Pore Size, Cell Biolabs
Supplier: Cell Biolabs
Certificates
About this item
CytoSelect™ Cell Migration (Chemotaxis) Assays are ideal for determining the chemotactic properties of cells
With the CytoSelect™ Cell Migration Assays, no manual cell counting is required. Quantitation of cells is performed using a colorimetric or fluorometric dye in a 96-well plate reader. The 8 µm pore size is suitable for most cell types including epithelial cells, fibroblasts, and cancer cell lines. Kits are available in 24-well and 96-well formats.
These kits use a Boyden Chamber in which inserts containing a membrane with a defined pore size are nested in the wells of a cell culture plate. Cells are seeded above the membrane and are allowed to migrate toward a chemoattractant in the lower chamber.
Specifications
- Storage Temperature:4°C