Order Entry
United States
ContactUsLinkComponent
OxiSelect™ Cellular Antioxidant Assay Kit (Green Fluorescence), Cell Biolabs
OxiSelect™ Cellular Antioxidant Assay Kit (Green Fluorescence), Cell Biolabs
Catalog # 102964-976
Supplier:  Cell Biolabs
CAS Number:  
undefined
OxiSelect™ Cellular Antioxidant Assay Kit (Green Fluorescence), Cell Biolabs
Catalog # 102964-976
Supplier:  Cell Biolabs
Supplier Number:  STA-349
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Cellular Antioxidant Assay Kit
  • Size:
    192 Assays
  • Storage Temperature:
    4°C and –20°C
  • Cat. No.:
    102964-976
  • Method:
    Fluorometric
  • Supplier no.:
    STA-349

Specifications

About this item

The OxiSelect™ Cellular Antioxidant Assay Kit is a cell-based assay for measuring the activity of an exogenous antioxidant compound within a cellular environment.  Adherent cells are first cultured in a 96-well black fluorescence cell culture plate until confluent

Then the cells are pre-incubated with a cell-permeable DCFH-DA fluorescence probe dye and the bioflavonoid Quercetin, or the antioxidant sample being tested.  After a brief incubation, the cells are washed, and the reaction started by adding the Free Radical Initiator.  The Free Radical Initiator creates free radicals that convert the probe to highly fluorescent DCF.  The Quercetin inhibits the formation of free radicals, and thus DCF formation, in a concentration dependent manner.