Order Entry
United States
Orders LinkContactUsLinkComponent
RIPA Buffer, Cell Biolabs

RIPA Buffer, Cell Biolabs

Marketsource
Supplier:   Cell Biolabs
RIPA Buffer, Cell Biolabs
Supplier:  Cell Biolabs
Marketsource
Certificates
RIPA Buffer, Cell Biolabs

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Cell Biolabs' RIPA Buffer is suitable for the extraction of cytoplasmic, membrane, and nuclear proteins from cells or tissues

The buffer is suitable for a variety of downstream applications such as reporter assays, protein assays, immunoassays and protein purification. RIPA Buffer is available with or without a separate Protease Inhibitor Cocktail.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 102963-644
  • 102963-646
  • Storage Temperature:
  • Room Temperature and –20°C
  • Room Temperature and –20°C

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 102963-644
    RIPA Buffer with Protease Inhibitor Cocktail
    RIPA Buffer with Protease Inhibitor Cocktail
    20mL
    Log in for availability
    Log in for availability
    Each
    Each - $129.23
    $129.23
    Each
       $129.23
    Minimum order quantity is 1

    Supplier Number: AKR-190
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    102963-644
    Storage Temperature
    Room Temperature and –20°C
    Marketsource
  • 102963-646
    RIPA Buffer
    RIPA Buffer
    20mL
    Log in for availability
    Log in for availability
    Each
    Each - $104.62
    $104.62
    Each
       $104.62
    Minimum order quantity is 1

    Supplier Number: AKR-191
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    102963-646
    Storage Temperature
    Room Temperature and –20°C
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.