Order Entry
United States
ContactUsLinkComponent
RFP ELISA Kit, Cell Biolabs
RFP ELISA Kit, Cell Biolabs
  102963-628
 :  
undefined
RFP ELISA Kit, Cell Biolabs
  102963-628
 :  AKR-122
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Description:
    RFP ELISA Kit
  • Size:
    96 Assays
  • Detection Method:
    Colorimetric
  • Storage Temperature:
    4°C and –80°C
  • Sensitivity:
    150 pg/mL
  • Cat. No.:
    102963-628
  • Supplier no.:
    AKR-122

 

 

Cell Biolabs’ RFP ELISA Kit is an enzyme immunoassay developed for detection and quantitation of RFP or RFP fusion protein in cell or tissue samples

The quantity of RFP in an unknown sample is determined by comparing its absorbance with that of a known recombinant RFP standard curve. The kit has detection sensitivity limit of 150 pg/mL RFP.