Order Entry
United States
ContactUsLinkComponent
Rapid GST Inclusion Body Solubilization and Renaturation Kit, Cell Biolabs
Rapid GST Inclusion Body Solubilization and Renaturation Kit, Cell Biolabs
Catalog # 102963-618
Supplier:  Cell Biolabs
CAS Number:  
undefined
Rapid GST Inclusion Body Solubilization and Renaturation Kit, Cell Biolabs
Catalog # 102963-618
Supplier:  Cell Biolabs
Supplier Number:  AKR-110
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    GST Inclusion Body Solubilization Kit
  • Size:
    1 Kit
  • Environmentally Preferable:
  • Storage Temperature:
    RT
  • Cat. No.:
    102963-618
  • Supplier no.:
    AKR-110

Specifications

About this item

Recombinant proteins expressed in bacteria often form inclusion bodies, especially when they are expressed at high levels

The Rapid GST Inclusion Body Solubilization and Renaturation Kit is designed to retrieve expressed GST fusion proteins in soluble form after lysis and extraction procedures. No lengthy dialysis or dilution steps are required, and no pH variation or redox pair are involved. Each kit contains sufficient reagents to solubilize and renature up to 5-10 liters of bacterial culture.