Orders LinkContactUsLinkComponent
PhosphoBlocker™ Blocking Reagent, Cell Biolabs
PhosphoBlocker™ Blocking Reagent, Cell Biolabs
Marketsource
Supplier:   Cell Biolabs
PhosphoBlocker™ Blocking Reagent, Cell Biolabs
Supplier:  Cell Biolabs
Marketsource
Certificates
PhosphoBlocker™ Blocking Reagent, Cell Biolabs
PhosphoBlocker™ Blocking Reagent, Cell Biolabs
2 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
PhosphoBLOCKER™ Blocking Reagent provides superior blocking by maximizing signal-to-noise ratio
The PhosphoBLOCKER™ reagent is an easy-to-use premixed dry blend that enhances low level phosphoprotein signal without increasing background. The reagent particluarly excels with very low levels of endogenous phopsphoproteins.
Most commercially available Western blot blockers, such as dry milk or serum, are sufficient to block unreactive sites on the membrane. However, they are not designed to preserve phosphoprotein antigens during blotting.
Documents
Specifications for Product Family
Specifications
Specifications
  • Catalog No:
  • 102963-610
  • 102963-612
  • Storage Temperature:
  • RT
  • RT
Specifications
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 102963-610
    PhosphoBlocker™ Reagent
    PhosphoBlocker™ Reagent
    1L
    Log in for availability
    Log in for availability
    Each
    Each - $338.46
    $338.46
    Each
       $338.46
    Minimum order quantity is 1
    Supplier Number: AKR-103
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    102963-610
    Storage Temperature
    RT
    Marketsource
  • 102963-612
    PhosphoBlocker™ Reagent
    PhosphoBlocker™ Reagent
    4L
    Log in for availability
    Log in for availability
    Each
    Each - $806.15
    $806.15
    Each
       $806.15
    Minimum order quantity is 1
    Supplier Number: AKR-104
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    102963-612
    Storage Temperature
    RT
    Marketsource