Tissue-Tek® FormaGO® Formalin Neutralizer, Sakura® Finetek
Catalog # 10852-346
Supplier: Sakura Finetek
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:FormaGO® Formalin Neutralizer
- Size:16/case
- Cat. No.:10852-346
- Packaging:Pouch
- Supplier no.:9151
Specifications
About this item
Each pre-measured pouch of Tissue-Tek® FormaGO® Formalin Neutralizer neutralizes one gallon of 10% formalin in approximately 5 minutes
There are no drain-clogging solids after neutralizing.