293AAV Cell Line, Cell Biolabs
Catalog # 102966-494
Supplier: Cell Biolabs
CAS Number:
Specifications
- Cell Format:Cryopreserved
- Description:293AAV Cells
- Size:1 Vial
- Storage Conditions:–80°C or LN2
- Cat. No.:102966-494
- Supplier no.:AAV-100
Specifications
About this item
The 293AAV Cell Line is derived from the parental 293 cells but selected for attributes that increase AAV production, including firmer attachment and larger surface area
This allows the cells to produce higher levels of AAV compared to standard 293 cells.