OxiSelect™ Catalase Activity Assay Kits, Cell Biolabs
Supplier: Cell Biolabs
Certificates
About this item
Catalase is a ubiquitous enzyme that destroys hydrogen peroxides formed during oxidative stress
The OxiSelect™ Catalase Activity Assay Kit (Fluorometric) provides a 40-fold increase in sensitivity compared to our colorimetric assay. Each kit provides sufficient reagents to perform up to 500 assays in a 96-well microtiter plate. This includes blanks, catalase standards and unknown samples.
OxiSelect™ Catalase Activity Assay Kits measure catalase activity in less than one hour from a variety of samples including blood, cells and tissues. The colorimetric kit provides sufficient reagents to perform up to 100 assays in a 96-well microtiter plate, or 50 assays in microcentrifuge tubes. This includes blanks, catalase standards and unknown samples.
Specifications
- Storage Temperature:4°C and –20°C