Order Entry
United States
ContactUsLinkComponent
OxiSelect™ 8-iso-Prostaglandin F2a ELISA Kit, Cell Biolabs
OxiSelect™ 8-iso-Prostaglandin F2a ELISA Kit, Cell Biolabs
Supplier:  Cell Biolabs
Certificates
undefined
OxiSelect™ 8-iso-Prostaglandin F2a ELISA Kit, Cell Biolabs
Supplier:  Cell Biolabs

Specifications

  • Storage Conditions:
    4°C and –20°C
  • Detection Method:
    Colorimetric
  • Sensitivity:
    49 pg/mL

Specifications

Related Information

Frequently Bought Together