About this item
OxiSelect™ TBARS Assay Kit provides a much more user-friendly protocol to measure the MDA-TBA adduct
Important Note: MDA adducts are not stable long term. For best results test all samples immediately upon collection, or freeze them at -80ºC for up to one month. MDA may be degraded in samples that have been frozen for longer periods; in such cases more reliable results may be obtained from more stable markers of oxidative stress such as protein carbonyl, 8-OHdG or 4-HNE.
The TBARS (Thiobarbituric Acid Reactive Substances) assay is well-established for screening and monitoring lipid peroxidation. MDA forms a 1:2 adduct with thiobarbituric acid; the MDA-TBA adduct can then be measured. Reaction volumes are much smaller than the traditional assay, so much less sample is required. Also, reactions can be performed in standard polypropylene tubes - no glass tubes or glass marbles are required. The assay provides a quick 30 minute assessment for oxidative stress.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Storage Temperature:4°C