Order Entry
United States
ContactUsLinkComponent
Recombinant Protein L (from E. coli)
Recombinant Protein L (from E. coli)
Supplier:  Avantor
Certificates
undefined
Recombinant Protein L (from E. coli)
Supplier:  Avantor

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Endotoxin Content:
    <0.1 ng/μg
  • Reconstitution Instructions:
    Reconstitution in H₂O to a concentration of 5 mg/ml gives a clear solution.
  • Protein Synonyms:
    r-protein L|IgG purification|protein L|protein/L|protein-L|rProtein L|Proteinl|Protein L
  • Protein/Peptide Name:
    Protein L
  • Purity:
    >98%
  • Molecular Weight:
    43.2 kDa
  • Formulation:
    Lyophilized with no additives

Specifications

Related Information

Frequently Bought Together