Order Entry
United States
ContactUsLinkComponent
Recombinant Turbo3C Hrv3C Protease (from E. coli)
Recombinant Turbo3C Hrv3C Protease (from E. coli)
Catalog # 10013-240
Supplier:  Avantor
CAS Number:  
undefined
Recombinant Turbo3C Hrv3C Protease (from E. coli)
Catalog # 10013-240
Supplier:  Avantor
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Enzyme type:
    Recombinant
  • Source:
    E. coli
  • Size:
    1 mg
  • Enzyme Name:
    Turbo3C Hrv3C Protease
  • Enzyme Synonyms:
    HRV3C Protease
  • Molecular Weight:
    47.0 kDa
  • Formulation:
    2 mg/ml in 25 mM Tris-HCl, pH 8.0, 50 mM NaCl, 1 mM TCEP, and 50% glycerol
  • Cat. No.:
    10013-240
  • Supplier no.:
    9206-1

Specifications

About this item

Pure recombinant active bacterial Turbo3C (HRV3C) Protease

BioVision’s Human rhinovirus 3C protease (HRV3C Protease) is a cysteine protease that recognizes the cleavage site of Leu-Glu-Val-Leu-Phe-Gln-Gly-Pro, commonly referred to as the PreScission Site. It cleaves between Gln and Gly (independent of Pro). The recombinant form of the HRV3C protease is a restriction grade protease that has robust activity at 4°C with high specific activity and great stability. It does not require any special buffer for its activity and can be used in a buffer most suitable for the target protein. This HRV3C Protease is a 47 kDa protein with both GST and His tags so it can be easily removed by either Ni-chelating or Glutathione (GSH) resin along with the cleaved tag.