Anti-FADS3 Rabbit Polyclonal Antibody
Catalog # 89517-020
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Fatty Acid Desaturase 3
- Antigen Symbol:FADS3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:C-Terminal
- Gene ID:3995
- Antigen Synonyms:CYB5RP|LLCDL3
- Amino Acid Number:328 to 355
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:51.145 kDa
- Sequence:HIPKEIGHEKHRDC
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping Temperature:4 °C
- Immunogen:This FADS3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 328-355 amino acids from the C-terminal region of human FADS3.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89517-020
- Supplier no.:AP18268B
Specifications
About this item
Western Blot: 1:1000
Type: Primary
Antigen: FADS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human