Anti-RBM7 Rabbit Polyclonal Antibody
89514-718
: Abgent
- Antibody Type:Primary
- Antigen Name:RNA Binding Motif Protein 7
- Antigen Symbol:RBM7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:400 µL
- Epitope:C-Terminal
- Gene ID:10179
- Amino Acid Number:196 to 222
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:30.504 kDa
- Sequence:RHYSREQRYTDHC
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping Temperature:4 °C
- Immunogen:This RBM7 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 196-222 amino acids from the C-terminal region of human RBM7.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89514-718
- Supplier no.:AP19532B
Western Blot: 1:1000
Type: Primary
Antigen: RBM7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human, Mouse