2-Propanol ≥99.8% (by GC), VWR Chemicals BDH®
Catalog # BDH11744LP
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Pack type:Poly bottle
- Size:4 L
- Cat. No.:BDH11744LP
Specifications
Specification Test Results
- Identification by FTIR:Equivalent to standard
- Appearance:Clear, colorless liquid
- Water (K.F.):Less than 0.050 %
- Refractive index (20 °C):1.3772 ± 0.0010
- Nonvolatile Residue:Less than 5 mg/l (0.0005 g/100 ml)
- Assay:Greater than 99.8 %
- Specific Gravity (25 °C):0.783 - 0.787
- Color (APHA):Less than 10
- Acidity:Passes test
Specification Test Results
About this item
:
:60.1 g/mol
Density:0.8…0.92 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-661-7
UN:1987
ADR:3,II
:GHS02
:GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674