Specifications
- Description:Hemastix Urinalysis Reagent Test Strips
- Environmentally Preferable:
- Cat. No.:10789-334
- Parameters tested:Blood
- Supplier no.:2190
Specifications
About this item
The Hemastix® test strip is the reliable frontline test for detection of blood.
- Reliable
- Detects blood in urine
- Ready to use
Hemastix® Reagent Test Strips from Siemens Healthcare Diagnostics are for professional, in vitro diagnostic use only. Hemastix® urinalysis test strips are ready to use upon removal from the bottle and may be read visually or read instrumentally using the CLINITEK Status®+ Urine Analyser.