2,2'-Dimethyl-2,2'-azodipropiononitrile ≥98%
Catalog # 101184-722
Supplier: Pfaltz & Bauer
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Size:25 g
- Environmentally Preferable:
- Cat. No.:101184-722
- Supplier no.:A37050
Specifications
Specification Test Results
- Chemical Name:2,2'-Azobis-2-methylpropionitrile 98%
- Synonyms:a,a'-Azoisobutyronitrile; AIBN; Azobisisobutyronitrile; VAZO® catalyst 64 free radical source
- Molecular Formula:(CH₃)₂C(CN)N₂C(CH₃)₂CN
- Molecular Weight:164.21
- Sensitivity:Air and Light sensitive
- Storage Temperature:2 to 8 ºC
- Appearance:White crystalline powder
- Melting Point:102-104 ºC
- Density:1.1 g/cm³
- Stability:Stable under normal temperatures and pressures
- Incompatibility:Aldehydes, Strong oxidizing agents, Alcohols, Alkali metals, Heptane, Contamination, Acetone
- Reactivity:React with incompatible materials to release other hazardous substances
- Conditions to Avoid:Heat, flame, sparks, other ignition sources
- Hazardous Decomposition Products:Carbon oxides, Nitrogen oxides
Specification Test Results
About this item
Synonyms: 2,2'-Azobis(2-methylpropionitril), α,α'-Azoisobutyronitrile, AIBN, ADZN, 2,2'-Azobisisobutyronitrile, 2,2'-Azobis(2-methylpropionitrile), Azoisobutyronitrile
:
:164.21
UN:3234
ADR:4.1,
CAS Number:78-67-1
Formula:C₈H₁₂N₄
MDL Number:MFCD00013808