Loading Pipette Tips for DispoEquilibrium Dialyzer, Harvard Apparatus
Catalog # 101167-460
Supplier: Harvard Apparatus
Specifications
- Packed:100/Pack
- Cat. No.:101167-460
- Tip style:Extra Loading Pippette Tips
- For use with:DispoEquilibrium Dialyzer
- Supplier no.:742222
Specifications
About this item
The QuikPrep® DispoEquilibrium Dialyzer™ made of inert PTFE is suitable for binding and interaction studies. It allows maximum sample retention.This is useful for studies which use radiolabeled compounds.
- Easy to use
- Disposable
- Small sample volumes: 25 to 75 μl each chamber
- Rapid dialysis due to ultra-thin membrane
- Leak-proof
High-quality regenerated cellulose membranes with MWCOs of 5000 and 10000 Daltons.