Dissection Tray, AIMS™
Dissection Tray, 9x6.5x1"
Catalog # 10060-188
Supplier: Aims
Supplier #: DTW1
Jump to:
Product Details & Documents
Blue wax dissection tray is formulated to provide proper consistency to allow pins to stick firmly and be easily removed.
- Uses a completely environmentally safe wax product
- Surface can be renewed by heating and the dissection tray can be reused several times
The MSDS sheet for this product lists no toxic chemicals and there are no fire, health or reactivity hazards with this product.
Specifications
- Description:Dissection Tray
- Size:9x6.5x1"