About this item
Purified anti-His Tag [J099B12]; Isotype: Mouse IgG1, κ; Reactivity: All species (6x His tag epitope); Apps: WB, ICC, ICFC; Size: 1 mg
Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.1 - 0.5 μg per ml (Dilution 1:1000 - 1:5000). For immunocytochemistry, a concentration range of 0.1 - 0.5 μg/ml (Dilution 1:1000 - 1:5000) is recommended. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: His Tag
Clonality: Monoclonal
Clone: J099B12
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: All species (6x His tag epitope)
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 10020-552
- 652501-BL
- 652502-BL
- Antigen Symbol:
- His Tag
- His Tag
- His Tag
- Antigen Synonyms:
- Histidine Tag epitope tag
- HIS|Histidine Tag epitope tag
- HIS|Histidine Tag epitope tag
- Antibody Type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Clone:
- J099B12
- J099B12
- J099B12
- Reactivity:
- Multiple
- Multiple
- Multiple
- Host:
- Mouse
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- IgG1 kappa
- Western Blot:
- Yes
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Yes
- Immunogen:
- KLH-conjugated His-tag peptide
- KLH-conjugated His-tag peptide
- KLH-conjugated His-tag peptide
- Form:
- This antibody is provided in phosphate-buffered solution, pH 7.2, 0.09% NaN3.
- This antibody is provided in phosphate-buffered solution, pH 7.2, 0.09% NaN3.
- This antibody is provided in phosphate-buffered solution, pH 7.2, 0.09% NaN3.
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 0.50 mg/mL
- 0.50 mg/mL
- 0.50 mg/mL
- Storage Temperature:
- Upon receipt, store undiluted between 2 °C and 8 °C.
- Upon receipt, store undiluted between 2 °C and 8 °C.
- Upon receipt, store undiluted between 2 °C and 8 °C.
- Shipping Temperature:
- Ambient RT
- Ambient RT
- Ambient RT