Anti-CD73 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: AD2]
Supplier: Biolegend
Certificates
About this item
PE/Dazzle™ 594 anti-human CD73 (Ecto-5'-nucleotidase) [AD2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD73
Clonality: Monoclonal
Clone: AD2
Conjugation: PE (Phycoerythrin)/Dazzle 594
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon
Specifications
- Catalog No:
- 10021-052
- 10021-050
- Antigen Symbol:
- CD73
- CD73
- Antigen Synonyms:
- NT5E|5’-NT|Ecto-5’-nucleotidase|L-VAP-2|E.C3.1.3.5
- NT5E|5’-NT|Ecto-5’-nucleotidase|L-VAP-2|E.C3.1.3.5
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)/Dazzle 594
- PE (Phycoerythrin)/Dazzle 594
- Clone:
- AD2
- AD2
- Reactivity:
- Baboon, African green monkey, Human
- Baboon, African green monkey, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 4907
- 4907
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PE/Dazzle™ 594 under optimal conditions. The solution is free of unconjugated PE/Dazzle™ 594 and unconjugated antibody.
- The antibody was purified by affinity chromatography and conjugated with PE/Dazzle™ 594 under optimal conditions. The solution is free of unconjugated PE/Dazzle™ 594 and unconjugated antibody.
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Specifications
Frequently Bought Together