Order Entry
United States
ContactUsLinkComponent
 
Anti-SIRPG Rabbit Polyclonal Antibody
  89519-622
 :  Abgent
Anti-SIRPG Rabbit Polyclonal Antibody
  89519-622
 :  Abgent
 :  AP16636C

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Signal Regulatory Protein Gamma
  • Antigen Symbol:
    SIRPG
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    400 µL
  • Epitope:
    Center
  • Gene ID:
    55423
  • Antigen Synonyms:
    SIRPB2|bA77C3.1|CD172g|SIRPgamma|SIRP-B2
  • Amino Acid Number:
    193 to 221
  • Storage Buffer:
    PBS with 0.09% (W/V) sodium azide, pH7.4
  • Molecular Weight:
    42.498 kDa
  • Sequence:
    RSTARVVLDPWDVRC
  • Storage Temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.48mg/ml
  • Shipping Temperature:
    4 °C
  • Immunogen:
    This SIRPG antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 193-221 amino acids from the Central region of human SIRPG.
  • Purification:
    Protein A & Peptide affinity purification
  • Cat. No.:
    89519-622
  • Supplier no.:
    AP16636C

 

 

Western Blot: 1:1000

Type: Primary
Antigen: SIRPG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Human