Order Entry
United States
ContactUsLinkComponent
Anti-XCR1 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: ZET]
Anti-XCR1 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: ZET]
Supplier:  Biolegend
Certificates
Anti-XCR1 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®) [clone: ZET]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • 10756-646
  • 10756-648
  • Antigen Symbol:
  • XCR1
  • XCR1
  • Antigen Synonyms:
  • XC chemokine receptor 1|CCXCR1|GPR5|mXCR1|Lymphotactin receptor|SCM1 receptor
  • XC chemokine receptor 1|CCXCR1|GPR5|mXCR1|Lymphotactin receptor|SCM1 receptor
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
  • PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
  • Clone:
  • ZET
  • ZET
  • Reactivity:
  • Rat, Mouse
  • Rat, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 23832, 301086
  • 23832, 301086
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with PerCP/Cy5.5 under optimal conditions. The solution is free of unconjugated PerCP/Cy5.5 and unconjugated antibody.
  • The antibody was purified by affinity chromatography and conjugated with PerCP/Cy5.5 under optimal conditions. The solution is free of unconjugated PerCP/Cy5.5 and unconjugated antibody.
  • Concentration:
  • 0.20 mg/mL
  • 0.20 mg/mL
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

Specifications

Frequently Bought Together