Anti-MOCS1 Rabbit Polyclonal Antibody
89515-186
: Abgent
- Antibody Type:Primary
- Antigen Name:Molybdenum Cofactor Synthesis 1
- Antigen Symbol:MOCS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:N-Terminal
- Gene ID:4337
- Antigen Synonyms:MOCOD|MIG11|MOCODA
- Amino Acid Number:31 to 58
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:70.105 kDa
- Sequence:RRQFLREHAAPFSC
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5mg/ml
- Shipping Temperature:4 °C
- Immunogen:This MOCS1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 31-58 amino acids from the N-terminal region of human MOCS1.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89515-186
- Supplier no.:AP19150A
Western Blot: 1:1000
Type: Primary
Antigen: MOCS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype:
Reactivity: Human