Order Entry
United States
ContactUsLinkComponent
Anti-TAS2R13 Rabbit Polyclonal Antibody
Anti-TAS2R13 Rabbit Polyclonal Antibody
Catalog # 89518-376
Supplier:  Abgent
Anti-TAS2R13 Rabbit Polyclonal Antibody
Catalog # 89518-376
Supplier:  Abgent
Supplier Number:  AP17339B

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Taste Receptor Type 2 Member 13
  • Antigen Symbol:
    TAS2R13
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    400 µL
  • Epitope:
    C-Terminal
  • Gene ID:
    50838
  • Antigen Synonyms:
    T2R13|TRB3
  • Amino Acid Number:
    199 to 228
  • Storage Buffer:
    PBS with 0.09% (W/V) sodium azide, pH7.4
  • Molecular Weight:
    35.118 kDa
  • Sequence:
    LNYKGHRDPRTKVHTC
  • Storage Temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.35mg/ml
  • Shipping Temperature:
    4 °C
  • Immunogen:
    This TAS2R13 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 199-228 amino acids from the C-terminal region of human TAS2R13.
  • Purification:
    Protein A & Peptide affinity purification
  • Cat. No.:
    89518-376
  • Supplier no.:
    AP17339B

Specifications

About this item

  • Western Blot: 1:1000

Type: Primary
Antigen: TAS2R13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype:
Reactivity: Human