Order Entry
United States
ContactUsLinkComponent
Anti-Podoplanin Rat Monoclonal Antibody (Biotin) [clone: NC-08]
Anti-Podoplanin Rat Monoclonal Antibody (Biotin) [clone: NC-08]
Catalog # 10758-560
Supplier:  Biolegend
Anti-Podoplanin Rat Monoclonal Antibody (Biotin) [clone: NC-08]
Catalog # 10758-560
Supplier:  Biolegend
Supplier Number:  337015

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Podoplanin
  • Clonality:
    Monoclonal
  • Clone:
    NC-08
  • Conjugation:
    Biotin
  • Host:
    Rat
  • Isotype:
    IgG2a lambda
  • Reactivity:
    Human
  • Size:
    50 μg
  • Form:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Gene ID:
    10630
  • Antigen Synonyms:
    stromal cell marker|PDPN|Aggrus|T1a|gp38
  • Storage Temperature:
    The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Concentration:
    0.50 mg/mL
  • Shipping Temperature:
    Ambient RT
  • Purification:
    The antibody was purified by affinity chromatography and conjugated with biotin under optimal conditions. The solution is free of unconjugated biotin.
  • Cat. No.:
    10758-560
  • Supplier no.:
    337015

Specifications

About this item

Biotin anti-human Podoplanin [NC-08]; Isotype: Rat IgG2a, λ; Reactivity: Human; Apps: FC; Size: 50 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: Podoplanin
Clonality: Monoclonal
Clone: NC-08
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: Rat IgG2a, λ
Reactivity: Human