Anti-Ly-6D Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 49-H4]
Supplier: Biolegend
Certificates
About this item
FITC anti-mouse Ly-6D [49-H4]; Isotype: Rat IgG2c, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: Ly-6D
Clonality: Monoclonal
Clone: 49-H4
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG2c, κ
Reactivity: Mouse
Specifications
- Catalog No:
- 138606-BL
- 138605-BL
- Antigen Symbol:
- Ly-6D
- Ly-6D
- Antigen Synonyms:
- ThB|Lymphocyte antigen 6D|LY6D|Thymocyte B-cell antigen
- ThB|Lymphocyte antigen 6D|LY6D|Thymocyte B-cell antigen
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- 49-H4
- 49-H4
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 17068
- 17068
- Isotype:
- IgG2c kappa
- IgG2c kappa
- Immunogen:
- BALB/c mouse plasmacytoma MOPC-104E
- BALB/c mouse plasmacytoma MOPC-104E
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC.
- The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC.
- Concentration:
- 0.50 mg/mL
- 0.50 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Specifications
Frequently Bought Together