Anti-IgG3, k Isotype Ctrl Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: MG3-35]
PE Mouse IgG3, κ Isotype Ctrl [MG3-35]; Isotype: Mouse IgG3, κ; Reactivity: Mouse; Apps: FC, ICFC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, use the isotype control at the same concentration as your primary antibody. Use our Concentration Lookup tool to find the exact concentrations of your lots of product.
Type: Control
Antigen: IgG3, k Isotype Ctrl
Clonality: Monoclonal
Clone: MG3-35
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG3, κ
Reactivity: Mouse
- Catalog No:
- 401320-BL
- 401319-BL
- Antigen Symbol:
- IgG3 k isotype ctrl
- IgG3 k isotype ctrl
- Antibody Type:
- Control
- Control
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- MG3-35
- MG3-35
- Reactivity:
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Isotype:
- IgG3 kappa
- IgG3 kappa
- Immunogen:
- Trinitrophenol + KLH
- Trinitrophenol + KLH
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- The antibody was purified by affinity chromatography, and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- Concentration:
- 0.20 mg/mL
- 0.20 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Frequently Bought Together