Anti-HLA-DM Mouse Monoclonal Antibody [clone: MaP.DM1]
Catalog # 10759-622
Supplier: Biolegend
Specifications
- Antibody Type:Primary
- Antigen Symbol:HLA-DM
- Clonality:Monoclonal
- Clone:MaP.DM1
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 μg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Antigen Synonyms:RING6|HLA DM|DMB|DMA,RING7|HLADM
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Concentration:0.50 mg/mL
- Shipping Temperature:Ambient RT
- Immunogen:MHC class II enriched organelles from the B cell line Pala.
- Purification:The antibody was purified by affinity chromatography.
- Cat. No.:10759-622
- Supplier no.:358002
Specifications
About this item
Purified anti-human HLA-DM [MaP.DM1]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC; Size: 100 μg
Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: HLA-DM
Clonality: Monoclonal
Clone: MaP.DM1
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human