Anti-GITR Ligand Rat Monoclonal Antibody [clone: 5F1]
Catalog # 10756-588
Supplier: Biolegend
Specifications
- Antibody Type:Primary
- Antigen Symbol:GITR Ligand
- Clonality:Monoclonal
- Clone:5F1
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Size:100 μg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Gene ID:240873
- Antigen Synonyms:TL6|AITRL|GITRL|TNFSF18
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Concentration:0.50 mg/mL
- Shipping Temperature:Ambient RT
- Purification:The antibody was purified by affinity chromatography.
- Cat. No.:10756-588
- Supplier no.:147402
Specifications
About this item
Purified anti-mouse GITR Ligand [5F1]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: GITR Ligand
Clonality: Monoclonal
Clone: 5F1
Conjugation:
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse