Order Entry
United States
ContactUsLinkComponent
Anti-GARP Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: F011-5]
Anti-GARP Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: F011-5]
Supplier:  Biolegend
Certificates
Anti-GARP Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: F011-5]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • 142904-BL
  • 142903-BL
  • Antigen Symbol:
  • GARP
  • GARP
  • Antigen Synonyms:
  • LRRC32|Glycoprotein A repetitions predominant|Garpin
  • LRRC32|Glycoprotein A repetitions predominant|Garpin
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • F011-5
  • F011-5
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 434215
  • 434215
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Immunogen:
  • Mouse GARP (extracellular domain)
  • Mouse GARP (extracellular domain)
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
  • The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
  • Concentration:
  • 0.20 mg/mL
  • 0.20 mg/mL
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

Specifications

Frequently Bought Together