Anti-Fc epsilon RI alpha Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: MAR-1]
Supplier: Biolegend
Certificates
About this item
PE/Cy7 anti-mouse FcεRIα [MAR-1]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: Fc epsilon RI alpha
Clonality: Monoclonal
Clone: MAR-1
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 134318-BL
- 134317-BL
- Antigen Symbol:
- Fc epsilon RI alpha
- Fc epsilon RI alpha
- Antigen Synonyms:
- FceRIa|FceRI-α|high affinity IgE receptormar1BasophilEosinophilMast CellPlateletsFcεRIα|FCER1A
- FceRIa|FceRI-α|high affinity IgE receptormar1BasophilEosinophilMast CellPlateletsFcεRIα|FCER1A
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)/Cy7®
- PE (Phycoerythrin)/Cy7®
- Clone:
- MAR-1
- MAR-1
- Reactivity:
- Mouse
- Mouse
- Host:
- Armenian hamster
- Armenian hamster
- Gene ID:
- 2205
- 2205
- Isotype:
- IgG
- IgG
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PE/Cy7 under optimal conditions. The solution is free of unconjugated PE/Cy7 and unconjugated antibody.
- The antibody was purified by affinity chromatography and conjugated with PE/Cy7 under optimal conditions. The solution is free of unconjugated PE/Cy7 and unconjugated antibody.
- Concentration:
- 0.20 mg/mL
- 0.20 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Specifications
Frequently Bought Together