Anti-Erythroid Cells Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: OX-83]
Supplier: Biolegend
Certificates
About this item
PE anti-rat Erythroid Cells [OX-83]; Isotype: Mouse IgG1; Reactivity: Rat; Apps: FC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: Erythroid Cells
Clonality: Monoclonal
Clone: OX-83
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat
Specifications
- Catalog No:
- 10756-972
- 10756-970
- Antigen Name:
- Erythroid Cells
- Erythroid Cells
- Antigen Synonyms:
- RBC|Rat erythrocytesMRC OX-83|OX83
- RBC|Rat erythrocytesMRC OX-83|OX83
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- OX-83
- OX-83
- Reactivity:
- Rat
- Rat
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
- Concentration:
- 0.20 mg/mL
- 0.20 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Specifications
Frequently Bought Together