Anti-Erythroid Cells Mouse Monoclonal Antibody [clone: OX-83]
Catalog # 10756-968
Supplier: Biolegend
Specifications
- Antibody Type:Primary
- Antigen Name:Erythroid Cells
- Clonality:Monoclonal
- Clone:OX-83
- Host:Mouse
- Isotype:IgG1
- Reactivity:Rat
- Size:100 μg
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Antigen Synonyms:RBC|Rat erythrocytesMRC OX-83|OX83
- Storage Temperature:The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Concentration:0.50 mg/mL
- Shipping Temperature:Ambient RT
- Purification:The antibody was purified by affinity chromatography.
- Cat. No.:10756-968
- Supplier no.:250402
Specifications
About this item
Purified anti-rat Erythroid Cells [OX-83]; Isotype: Mouse IgG1; Reactivity: Rat; Apps: FC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: Erythroid Cells
Clonality: Monoclonal
Clone: OX-83
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Rat