Anti-SDF4 Rabbit Polyclonal Antibody
Catalog # 89518-212
Supplier: Abgent
Specifications
- Antibody Type:Primary
- Antigen Name:Stromal Cell Derived Factor 4
- Antigen Symbol:SDF4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:400 µL
- Epitope:C-Terminal
- Gene ID:51150
- Antigen Synonyms:45 kDa calcium-binding protein|Cab45|PSEC0034|Stromal cell-derived factor 4|SDF4|SDF-4
- Amino Acid Number:258 to 287
- Storage Buffer:PBS with 0.09% (W/V) sodium azide, pH7.4
- Molecular Weight:41.807 kDa
- Sequence:DIDDNWVKDRKKEFEC
- Storage Temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.45mg/ml
- Shipping Temperature:4 °C
- Immunogen:This SDF4 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 258-287 amino acids from the C-terminal region of human SDF4.
- Purification:Protein A & Peptide affinity purification
- Cat. No.:89518-212
- Supplier no.:AP17498B
Specifications
About this item
Western Blot: 1:1000
Type: Primary
Antigen: SDF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human