Anti-CD49b Mouse Monoclonal Antibody [clone: P1E6-C5]
Supplier: Biolegend
Certificates
About this item
Purified anti-human CD49b [P1E6-C5]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, IP, ELISA; Size: 500 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD49b
Clonality: Monoclonal
Clone: P1E6-C5
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Specifications
- Catalog No:
- 10760-566
- 10760-564
- Antigen Symbol:
- CD49b
- CD49b
- Antigen Synonyms:
- integrin a2 chain|VLA-2 α chain|Integrin α2 chain alpha2 integrin|α2 integrin|alpha 2 integrin|ITGA2
- integrin a2 chain|VLA-2 α chain|Integrin α2 chain alpha2 integrin|α2 integrin|alpha 2 integrin|ITGA2
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- P1E6-C5
- P1E6-C5
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 3673
- 3673
- Isotype:
- IgG1 kappa
- IgG1 kappa
- ELISA:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Gel Shift:
- Yes
- Yes
- Immunogen:
- HT1080 cells
- HT1080 cells
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography.
- The antibody was purified by affinity chromatography.
- Concentration:
- 0.50 mg/mL
- 0.50 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C.
- The antibody solution should be stored undiluted between 2 °C and 8 °C.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Specifications
Frequently Bought Together