Order Entry
United States
ContactUsLinkComponent
Anti-CD47 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: miap301]
Anti-CD47 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: miap301]
Supplier:  Biolegend
Certificates
Anti-CD47 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: miap301]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • 10757-080
  • 10757-082
  • Antigen Symbol:
  • CD47
  • CD47
  • Antigen Synonyms:
  • IAP|Integrin-associated protein|Rh-associated protein|neurophilin,gp42
  • IAP|Integrin-associated protein|Rh-associated protein|neurophilin,gp42
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • miap301
  • miap301
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 16423
  • 16423
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
  • The antibody was purified by affinity chromatography and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
  • Concentration:
  • 0.20 mg/mL
  • 0.20 mg/mL
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

Specifications

Frequently Bought Together