Anti-CD43 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: S11]
Supplier: Biolegend
Certificates
About this item
FITC anti-mouse CD43 [S11]; Isotype: Rat IgG2b; Reactivity: Mouse; Apps: FC; Size: 200 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD43
Clonality: Monoclonal
Clone: S11
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgG2b
Reactivity: Mouse
Specifications
- Catalog No:
- 143204-BL
- 143203-BL
- Antigen Symbol:
- CD43
- CD43
- Antigen Synonyms:
- Sialophorin|Leukocyte Sialoglycoprotein|Leukosialin|SPN|Ly-48|gp115
- Sialophorin|Leukocyte Sialoglycoprotein|Leukosialin|SPN|Ly-48|gp115
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- S11
- S11
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 20737
- 20737
- Isotype:
- IgG2b
- IgG2b
- Immunogen:
- Mouse plasmacytoma cells
- Mouse plasmacytoma cells
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC.
- The antibody was purified by affinity chromatography, and conjugated with FITC under optimal conditions. The solution is free of unconjugated FITC.
- Concentration:
- 0.50 mg/mL
- 0.50 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Specifications
Frequently Bought Together