Order Entry
United States
ContactUsLinkComponent
 
Anti-CD40 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 5C3]
 :  Biolegend
 
Anti-CD40 Mouse Monoclonal Antibody (Alexa Fluor® 700) [clone: 5C3]
 :  Biolegend

 

  • Catalog No:
  • 10757-608
  • 10757-606
  • Antigen Symbol:
  • CD40
  • CD40
  • Antigen Synonyms:
  • BP50|TNFRSF5
  • BP50|TNFRSF5
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 700
  • Alexa Fluor® 700
  • Clone:
  • 5C3
  • 5C3
  • Reactivity:
  • Baboon, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Squirrel monkey
  • Baboon, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Squirrel monkey
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 958
  • 958
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 700 under optimal conditions.
  • The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 700 under optimal conditions.
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

 

Frequently Bought Together