Anti-CD3 epsilon Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: 145-2C11]
Supplier: Biolegend
Certificates
About this item
PE/Dazzle™ 594 anti-mouse CD3ε [145-2C11]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD3epsilon
Clonality: Monoclonal
Clone: 145-2C11
Conjugation: PE (Phycoerythrin)/Dazzle 594
Epitope:
Host: Armenian Hamster
Isotype: Armenian Hamster IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 10756-658
- 10756-656
- Antigen Symbol:
- CD3 epsilon
- CD3 epsilon
- Antigen Synonyms:
- T3|CD3|CD3ε|CD3epsilon|CD3 epsilon
- T3|CD3|CD3ε|CD3epsilon|CD3 epsilon
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)/Dazzle 594
- PE (Phycoerythrin)/Dazzle 594
- Clone:
- 145-2C11
- 145-2C11
- Reactivity:
- Mouse
- Mouse
- Host:
- Armenian hamster
- Armenian hamster
- Gene ID:
- 12501
- 12501
- Isotype:
- IgG
- IgG
- Immunogen:
- H-2Kb-specific mouse cytotoxic T lymphocyte clone BM10-37
- H-2Kb-specific mouse cytotoxic T lymphocyte clone BM10-37
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PE/Dazzle™ 594 under optimal conditions. The solution is free of unconjugated PE/Dazzle™ 594 and unconjugated antibody.
- The antibody was purified by affinity chromatography and conjugated with PE/Dazzle™ 594 under optimal conditions. The solution is free of unconjugated PE/Dazzle™ 594 and unconjugated antibody.
- Concentration:
- 0.20 mg/mL
- 0.20 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Specifications
Frequently Bought Together