Order Entry
United States
ContactUsLinkComponent
 
Anti-CD371 (CLEC12A) Mouse Monoclonal Antibody [clone: 50C1]
  353602-BL
 :  Biolegend
Anti-CD371 (CLEC12A) Mouse Monoclonal Antibody [clone: 50C1]
  353602-BL
 :  Biolegend
 :  353602
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

 

  • Antibody Type:
    Primary
  • Antigen Symbol:
    CD371 (CLEC12A)
  • Clonality:
    Monoclonal
  • Clone:
    50C1
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Size:
    100 μg
  • Form:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Gene ID:
    160364
  • Antigen Synonyms:
    CLL-1|MICL|CLEC12A|CD371|CLL1|DCAL-2|C-type lectin domain family 12 member A
  • Storage Temperature:
    The antibody solution should be stored undiluted between 2 °C and 8 °C.
  • Concentration:
    0.50 mg/mL
  • Shipping Temperature:
    Ambient RT
  • Immunogen:
    293T cells expressing CLEC12A-Flag
  • Purification:
    The antibody was purified by affinity chromatography.
  • Cat. No.:
    353602-BL
  • Supplier no.:
    353602

 

 

Purified anti-human CD371 (CLEC12A) [50C1]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.25 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD371 (CLEC12A)
Clonality: Monoclonal
Clone: 50C1
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human