Order Entry
United States
ContactUsLinkComponent
 
Anti-CD309/VEGFR2 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: 7D4-6]
 :  Biolegend
 
Anti-CD309/VEGFR2 Mouse Monoclonal Antibody (Alexa Fluor® 647) [clone: 7D4-6]
 :  Biolegend

 

  • Catalog No:
  • 10760-694
  • 10760-696
  • Antigen Symbol:
  • CD309/VEGFR2
  • CD309/VEGFR2
  • Antigen Synonyms:
  • KDR|VEGFR2)|Vascular Endothelial Receptor 2 (VEGF-R2
  • KDR|VEGFR2)|Vascular Endothelial Receptor 2 (VEGF-R2
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 647
  • Alexa Fluor® 647
  • Clone:
  • 7D4-6
  • 7D4-6
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 3791
  • 3791
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Immunogen:
  • Human KDR recombinant protein
  • Human KDR recombinant protein
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 647 under optimal conditions.
  • The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 647 under optimal conditions.
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

 

Frequently Bought Together