Order Entry
United States
ContactUsLinkComponent
Anti-CD298 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: LNH-94]
Anti-CD298 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: LNH-94]
Catalog # 10758-640
Supplier:  Biolegend
Anti-CD298 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: LNH-94]
Catalog # 10758-640
Supplier:  Biolegend
Supplier Number:  341706

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    CD298
  • Clonality:
    Monoclonal
  • Clone:
    LNH-94
  • Conjugation:
    APC (Allophycocyanin)
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Baboon,
    African green monkey,
    Cynomolgus monkey,
    Rhesus monkey,
    Human
  • Size:
    100 tests
  • Form:
    Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Gene ID:
    483
  • Antigen Synonyms:
    Na+/K+ ATPase β3|ATP1B3
  • Storage Temperature:
    The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
    Ambient RT
  • Purification:
    The antibody was purified by affinity chromatography and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
  • Cat. No.:
    10758-640
  • Supplier no.:
    341706

Specifications

About this item

APC anti-human CD298 [LNH-94]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD298
Clonality: Monoclonal
Clone: LNH-94
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus