Order Entry
United States
ContactUsLinkComponent
Anti-CD294 Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: BM16]
Anti-CD294 Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: BM16]
Supplier:  Biolegend
Certificates
Anti-CD294 Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: BM16]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • 350103-BL
  • 350104-BL
  • Antigen Symbol:
  • CD294
  • CD294
  • Antigen Synonyms:
  • GPR44|Chemoattractant receptor-homologous molecule expressed on Th2 cells|DP2|PTGDR2|G-protein coupled receptor 44
  • GPR44|Chemoattractant receptor-homologous molecule expressed on Th2 cells|DP2|PTGDR2|G-protein coupled receptor 44
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Alexa Fluor® 647
  • Alexa Fluor® 647
  • Clone:
  • BM16
  • BM16
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 11251
  • 11251
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Immunogen:
  • Rat cell line TART/B19-12.10 transfected with human CRTH2
  • Rat cell line TART/B19-12.10 transfected with human CRTH2
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Purification:
  • The antibody was purified by affinity chromatography, and conjugated with Alexa Fluor® 647 under optimal conditions.
  • The antibody was purified by affinity chromatography, and conjugated with Alexa Fluor® 647 under optimal conditions.
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

Specifications

Frequently Bought Together