Anti-CD274 (B7-H1, PD-L1) Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 29E.2A3]
Supplier: Biolegend
Certificates
About this item
PE/Cy7 anti-human CD274 (B7-H1, PD-L1) [29E.2A3]; Isotype: Mouse IgG2b, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD274 (B7-H1, PD-L1)
Clonality: Monoclonal
Clone: 29E.2A3
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Mouse
Isotype: Mouse IgG2b, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Specifications
- Catalog No:
- 10758-400
- 10758-398
- Antigen Symbol:
- CD274 (B7-H1, PD-L1)
- CD274 (B7-H1, PD-L1)
- Antigen Synonyms:
- PDL1|Programmed cell death ligand 1 (PD-L1)|29E2A3|B7 homolog 1 (B7-H1) B7H1|CD274|Immune checkpoints
- PDL1|Programmed cell death ligand 1 (PD-L1)|29E2A3|B7 homolog 1 (B7-H1) B7H1|CD274|Immune checkpoints
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)/Cy7®
- PE (Phycoerythrin)/Cy7®
- Clone:
- 29E.2A3
- 29E.2A3
- Reactivity:
- Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Human
- Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 29126
- 29126
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Immunogen:
- Full length human PD-L1
- Full length human PD-L1
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PE/Cy7 under optimal conditions. The solution is free of unconjugated PE/Cy7 and unconjugated antibody.
- The antibody was purified by affinity chromatography and conjugated with PE/Cy7 under optimal conditions. The solution is free of unconjugated PE/Cy7 and unconjugated antibody.
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Specifications
Frequently Bought Together