Order Entry
United States
ContactUsLinkComponent
Anti-CD269 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 19F2]
Anti-CD269 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 19F2]
Supplier:  Biolegend
Certificates
Anti-CD269 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 19F2]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • 10760-272
  • 10760-270
  • Antigen Symbol:
  • CD269
  • CD269
  • Antigen Synonyms:
  • TNFRSF17 B cell maturation antigen|TNFRSF17|BCM
  • TNFRSF17 B cell maturation antigen|TNFRSF17|BCM
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • 19F2
  • 19F2
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 608
  • 608
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Immunogen:
  • BCMA-mouse IgG Fc fusion protein
  • BCMA-mouse IgG Fc fusion protein
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
  • The antibody was purified by affinity chromatography and conjugated with PE under optimal conditions. The solution is free of unconjugated PE and unconjugated antibody.
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

Specifications

Frequently Bought Together