Anti-CD22 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: OX-97]
APC anti-mouse CD22 [OX-97]; Isotype: Rat IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg
Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.
Type: Primary
Antigen: CD22
Clonality: Monoclonal
Clone: OX-97
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse
- Catalog No:
- 10757-040
- 10757-042
- Antigen Symbol:
- CD22
- CD22
- Antigen Synonyms:
- BL-CAM|Lyb-8
- BL-CAM|Lyb-8
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- OX-97
- OX-97
- Reactivity:
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 12483
- 12483
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Form:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Purification:
- The antibody was purified by affinity chromatography and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
- The antibody was purified by affinity chromatography and conjugated with APC under optimal conditions. The solution is free of unconjugated APC and unconjugated antibody.
- Concentration:
- 0.20 mg/mL
- 0.20 mg/mL
- Storage Temperature:
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Shipping Temperature:
- Ambient RT
- Ambient RT
Frequently Bought Together