Order Entry
United States
ContactUsLinkComponent
Anti-CD206 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: C068C2]
Anti-CD206 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: C068C2]
Supplier:  Biolegend
Certificates
Anti-CD206 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: C068C2]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • 10757-252
  • 10757-250
  • Antigen Symbol:
  • CD206
  • CD206
  • Antigen Synonyms:
  • CD206|MMR (macrophage mannose receptor)|MRC1|MR (mannose receptor)
  • CD206|MMR (macrophage mannose receptor)|MRC1|MR (mannose receptor)
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)/Cy7®
  • PE (Phycoerythrin)/Cy7®
  • Clone:
  • C068C2
  • C068C2
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 17533
  • 17533
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Flow Cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant mouse CD206 (MMR)
  • Recombinant mouse CD206 (MMR)
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with PE/Cy7 under optimal conditions. The solution is free of unconjugated PE/Cy7 and unconjugated antibody.
  • The antibody was purified by affinity chromatography and conjugated with PE/Cy7 under optimal conditions. The solution is free of unconjugated PE/Cy7 and unconjugated antibody.
  • Concentration:
  • 0.20 mg/mL
  • 0.20 mg/mL
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

Specifications

Frequently Bought Together