Order Entry
United States
ContactUsLinkComponent
Anti-CD197 (CCR7) Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: G043H7]
Anti-CD197 (CCR7) Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: G043H7]
Supplier:  Biolegend
Certificates
Anti-CD197 (CCR7) Mouse Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: G043H7]
Supplier:  Biolegend

Specifications

  • Catalog No:
  • 353225-BL
  • 353226-BL
  • Antigen Symbol:
  • CD197 (CCR7)
  • CD197 (CCR7)
  • Antigen Synonyms:
  • CDw197|BLR2|CCR7
  • CDw197|BLR2|CCR7
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)/Cy7®
  • PE (Phycoerythrin)/Cy7®
  • Clone:
  • G043H7
  • G043H7
  • Reactivity:
  • Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Human
  • Baboon, African green monkey, Cynomolgus monkey, Rhesus monkey, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 1236
  • 1236
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Immunogen:
  • CCR7-transfected cells
  • CCR7-transfected cells
  • Form:
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
  • Purification:
  • The antibody was purified by affinity chromatography and conjugated with PE/Cy7 under optimal conditions. The solution is free of unconjugated PE/Cy7 and unconjugated antibody.
  • The antibody was purified by affinity chromatography and conjugated with PE/Cy7 under optimal conditions. The solution is free of unconjugated PE/Cy7 and unconjugated antibody.
  • Storage Temperature:
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • The antibody solution should be stored undiluted between 2 °C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
  • Shipping Temperature:
  • Ambient RT
  • Ambient RT

Specifications

Frequently Bought Together